1-Ethyl-1-(p-tolyl)hydrazine 95%
Lieferant: Apollo Scientific
Synonyme:
1-Ethyl-1-(4-methylphenyl)hydrazine
OR919938-10G
OR919938-1G
OR919938-5G
APOSOR919938-10GEA
596
EUR
APOSOR919938-10G
APOSOR919938-1G
APOSOR919938-5G
1-Ethyl-1-(p-tolyl)hydrazine 95%
1-Ethyl-1-(p-tolyl)hydrazine
Formel:
C₉H₁₄N₂ Molecular Weight: 150.23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03844793 CAS-Nummer: 61715-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...