Methyl-phenylcarbamat 95%
Lieferant: Apollo Scientific
Synonyme:
N-Phenylcarbamic acid Methyl Ester, Methyl N-Phenylcarbamate
OR919990-25G
OR919990-5G
OR919990-100G
APOSOR919990-25GEA
71.2
EUR
APOSOR919990-25G
APOSOR919990-5G
APOSOR919990-100G
Methyl-phenylcarbamat 95%
Methyl-phenylcarbamat
Formel:
C₈H₉NO₂ Molecular Weight: 151,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00027552 CAS-Nummer: 2603-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...