Methylhydrogenglutarat 95%
Lieferant: Apollo Scientific
Synonyme:
5-Methoxy-5-oxopentanoic acid
OR920036-25G
APOSOR920036-25GEA
60.9
EUR
APOSOR920036-25G
Methylhydrogenglutarat 95%
Methylhydrogenglutarat
Formel:
C₆H₁₀O₄ Molecular Weight: 146,1412 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004409 CAS-Nummer: 1501-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...