5-Phenyl-o-anisidin 95%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-4-phenylanisole, 2-Methoxy-5-phenylaniline, 5-Phenyl-o-anisidine, 3-Amino-4-methoxybiphenyl

OR920042-1G OR920042-25G OR920042-5G
APOSOR920042-1GEA 125 EUR
APOSOR920042-1G APOSOR920042-25G APOSOR920042-5G
5-Phenyl-o-anisidin 95%
5-Phenyl-o-anisidin
Formel: C₁₃H₁₃NO
Molecular Weight: 199,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007790
CAS-Nummer: 39811-17-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR