2-Hydroxyiminopropiophenon 98%

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxyiminopropiophenone, 2-Isonitrosopropiophenone, 1-Phenyl-1,2-propanedione-2-oxime, 1-Phenyl-1,2-propanedione 2-Oxime

OR920080-25G OR920080-5G
APOSOR920080-25GEA 219 EUR
APOSOR920080-25G APOSOR920080-5G
2-Hydroxyiminopropiophenon 98%
2-Hydroxyiminopropiophenon
Formel: C₉H₉NO₂
Molecular Weight: 163.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002115
CAS-Nummer: 119-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR