Allylphenylether 98%

Lieferant: Apollo Scientific

Synonyme: Phenylallylether

OR920084-100G OR920084-500G
APOSOR920084-100GEA 75.4 EUR
APOSOR920084-100G APOSOR920084-500G
Allylphenylether 98%
Allylphenylether
Formel: H₂CCHCH₂OC₆H₅
Molecular Weight: 134,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008644
CAS-Nummer: 1746-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR