(4-Methoxy-1-naphthyl)methanol 96%
Lieferant: Apollo Scientific
Synonyme:
1-(Hydroxymethyl)-4-methoxynaphthalene, (4-Methoxynaphthalen-1-yl)methanol
OR920087-5G
APOSOR920087-5GEA
194
EUR
APOSOR920087-5G
(4-Methoxy-1-naphthyl)methanol 96%
(4-Methoxy-1-naphthyl)methanol
Formel:
C₁₂H₁₂O₂ Molecular Weight: 188,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04089264 CAS-Nummer: 16820-54-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...