Vinyl methacrylate 98%
Lieferant: Apollo Scientific
Synonyme:
Methacrylic Acid Vinyl Ester
OR920125-25G
OR920125-5G
APOSOR920125-25GEA
79.9
EUR
APOSOR920125-25G
APOSOR920125-5G
Vinyl methacrylate 98%
Vinyl methacrylate
Formel:
C₆H₈O₂ Molecular Weight: 112,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059329 CAS-Nummer: 4245-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...