Methyl-1-methyl-2-pyrrolacetat 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(1-methyl-1H-pyrrol-2-yl)acetate, 1-Methylpyrrole-2-acetic Acid Methyl Ester

OR920190-25G OR920190-5G
APOSOR920190-25GEA 200 EUR
APOSOR920190-25G APOSOR920190-5G
Methyl-1-methyl-2-pyrrolacetat 95%
Methyl-1-methylpyrrole-2-acetate
Formel: C₈H₁₁NO₂
Molecular Weight: 153.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009752
CAS-Nummer: 51856-79-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR