6-Hydroxy-4-methylpyridin-2(1H)-one 95%
Lieferant: Apollo Scientific
Synonyme:
2,6-Dihydroxy-4-methylpyridine
OR920202-1G
OR920202-5G
APOSOR920202-1GEA
75.4
EUR
APOSOR920202-1G
APOSOR920202-5G
6-Hydroxy-4-methylpyridin-2(1H)-one 95%
6-Hydroxy-4-methylpyridin-2(1H)-one
Formel:
C₆H₇NO₂ Molecular Weight: 125,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17016132 CAS-Nummer: 4664-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...