(4-Pyridyl)essigsäureethylester 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl-(4-pyridyl)acetat

OR920243-25G OR920243-5G
APOSOR920243-25GEA 369 EUR
APOSOR920243-25G APOSOR920243-5G
(4-Pyridyl)essigsäureethylester 95%
(4-Pyridyl)essigsäureethylester
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051830
CAS-Nummer: 54401-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR