3-(Diethylamino)-1-propanol 95%

Lieferant: Apollo Scientific

Synonyme: 3-(Diethylamino)propan-1-ol, 3-Diethylamino-1-propanol

OR920298-100G
APOSOR920298-100GEA 149 EUR
APOSOR920298-100G
3-(Diethylamino)-1-propanol 95%
3-(Diethylamino)-1-propanol
Formel: C₇H₁₇NO
Molecular Weight: 131,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002947
CAS-Nummer: 622-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR