6-Hepten-1-ol 95%

Lieferant: Apollo Scientific

OR920322-1G OR920322-250MG OR920322-5G
APOSOR920322-1GEA 270 EUR
APOSOR920322-1G APOSOR920322-250MG APOSOR920322-5G
6-Hepten-1-ol 95%
6-Hepten-1-ol
Formel: C₇H₁₄O
Molecular Weight: 114,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00229167
CAS-Nummer: 4117-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR