1-Ethyl-3-pyrrolidinol 95%
Lieferant: Apollo Scientific
Synonyme:
1-Ethyl-3-hydroxypyrrolidine
OR920330-5G
APOSOR920330-5GEA
56.7
EUR
APOSOR920330-5G
1-Ethyl-3-pyrrolidinol 95%
1-Ethyl-3-pyrrolidinol
Formel:
C₆H₁₃NO Molecular Weight: 115,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003179 CAS-Nummer: 30727-14-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...