Butylcarbamat 95%

Lieferant: Apollo Scientific

Synonyme: Carbamidsäurebutylester

OR920407-100G OR920407-500G
APOSOR920407-100GEA 82.8 EUR
APOSOR920407-100G APOSOR920407-500G
Butylcarbamat 95%
Butylcarbamat
Formel: C₅H₁₁NO₂
Molecular Weight: 117,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007967
CAS-Nummer: 592-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR