3-Mercaptoisobutyric acid 95%
Lieferant: Apollo Scientific
Synonyme:
3-Mercapto-2-methylpropionic Acid
OR920443-1G
OR920443-25G
OR920443-5G
APOSOR920443-1GEA
132
EUR
APOSOR920443-1G
APOSOR920443-25G
APOSOR920443-5G
3-Mercaptoisobutyric acid 95%
3-Mercaptoisobutyric acid
Formel:
C₄H₈O₂S Molecular Weight: 120,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00671555 CAS-Nummer: 26473-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...