3-(Piperidin-1-ylmethyl)phenylboronsäurepinakolester 97%
Lieferant: Apollo Scientific
Synonyme:
3-(Piperidin-1-ylmethyl)phenylboronic acid, pinacol ester, 1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperidine
OR9206-1G
OR9206-250MG
APOSOR9206-1GEA
259
EUR
APOSOR9206-1G
APOSOR9206-250MG
3-(Piperidin-1-ylmethyl)phenylboronsäurepinakolester 97%
3-(Piperidin-1-ylmethyl)phenylboronsäurepinakolester
Formel:
C₁₈H₂₈BNO₂ Molecular Weight: 301.2314 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08435871 CAS-Nummer: 859833-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...