2-(4-(Methoxycarbonyl)phenyl)acetic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR920675-5G
APOSOR920675-5GEA
179
EUR
APOSOR920675-5G
2-(4-(Methoxycarbonyl)phenyl)acetic acid 98%
2-(4-(Methoxycarbonyl)phenyl)acetic acid
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194,1840 g/mol Schmelzpunkt: 112…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD19440718 CAS-Nummer: 22744-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...