3-Dimethylamino-2,2-dimethyl-1-propanol 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Dimethylaminoneopentanol
OR920692-100G
OR920692-25G
OR920692-5G
APOSOR920692-100GEA
539
EUR
APOSOR920692-100G
APOSOR920692-25G
APOSOR920692-5G
3-Dimethylamino-2,2-dimethyl-1-propanol 97%
3-Dimethylamino-2,2-dimethyl-1-propanol
Formel:
C₇H₁₇NO Molecular Weight: 131,22 g/mol Siedepunkt: 69 °C/15mmHg Dichte: 0,86 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059879 CAS-Nummer: 19059-68-8 EINECS: 242-791-7 UN: 2924 ADR: 3+8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...