Benzylchloracetat 98%
Lieferant: Apollo Scientific
Synonyme:
Chloressigsäurebenzylester, Phenylmethylchloracetat
OR920707-100G
OR920707-25G
APOSOR920707-100GEA
156
EUR
APOSOR920707-100G
APOSOR920707-25G
Benzylchloracetat 98%
Benzylchloracetat
Formel:
C₉H₉ClO₂ Molecular Weight: 184,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00157062 CAS-Nummer: 140-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...