3-Ethoxy-N,N-diethylanilin 95%
Lieferant: Apollo Scientific
Synonyme:
N,N-Diethyl-m-phenetidine
OR920744-25G
APOSOR920744-25GEA
105
EUR
APOSOR920744-25G
3-Ethoxy-N,N-diethylanilin 95%
3-Ethoxy-N,N-diethylanilin
Formel:
C₁₂H₁₉NO Molecular Weight: 193,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043936 CAS-Nummer: 1864-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...