(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine 95%
Lieferant: Apollo Scientific
OR920747-1G
APOSOR920747-1GEA
82.8
EUR
APOSOR920747-1G
(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine 95%
(S)-(+)-1-(2-Pyrrolidinylmethyl)pyrrolidine
Formel:
C₉H₁₈N₂ Molecular Weight: 154,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013441 CAS-Nummer: 51207-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...