4-Nitrosoresorcinol-1-monomethyl ether 95%

Lieferant: Apollo Scientific

Synonyme: Coniferron, 2-Hydroxy-4-methoxy-1-nitrosobenzene, 5-Methoxy-2-nitrosophenol, 4-Methoxy-1,2-benzoquinone 1-Monoxime

OR921045-1G OR921045-25G
APOSOR921045-1GEA 132 EUR
APOSOR921045-1G APOSOR921045-25G
4-Nitrosoresorcinol-1-monomethyl ether 95%
4-Nitrosoresorcinol-1-monomethyl ether
Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059952
CAS-Nummer: 13895-38-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR