3-Chlor-2,2-dimethyl-1-propanol 95%

Lieferant: Apollo Scientific

OR921073-25G OR921073-100G
APOSOR921073-25GEA 68.3 EUR
APOSOR921073-25G APOSOR921073-100G
3-Chlor-2,2-dimethyl-1-propanol 95%
3-Chlor-2,2-dimethyl-1-propanol
Formel: C₅H₁₁ClO
Molecular Weight: 122,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004684
CAS-Nummer: 13401-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR