Allyl-chloracetat 95%

Lieferant: Apollo Scientific

Synonyme: Chloroacetic acid allyl ester, Allyl 2-chloroacetate

OR921089-25G
APOSOR921089-25GEA 55.1 EUR
APOSOR921089-25G
Allyl-chloracetat 95%
Allyl-chloracetat
Formel: C₅H₇ClO₂
Molecular Weight: 134,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058936
CAS-Nummer: 2916-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR