(S)-(-)-N,N-Dimethyl-1-phenylethylamine 95%
Lieferant: Apollo Scientific
Synonyme:
(S)-(-)-alpha-Methyl-N,N-dimethylbenzylamine, (S)-N,N-Dimethyl-1-phenylethanamine
OR921111-5G
APOSOR921111-5GEA
260
EUR
APOSOR921111-5G
(S)-(-)-N,N-Dimethyl-1-phenylethylamine 95%
(S)-(-)-N,N-Dimethyl-1-phenylethylamine
Formel:
C₁₀H₁₅N Molecular Weight: 149,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00056236 CAS-Nummer: 17279-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...