Ethyl-2-cyanpropanoat 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-cyanopropanoate, 2-Cyanopropionic Acid Ethyl Ester

OR921115-25G OR921115-5G OR921115-100G
APOSOR921115-25GEA 190 EUR
APOSOR921115-25G APOSOR921115-5G APOSOR921115-100G
Ethyl-2-cyanpropanoat 97%
Ethyl-2-cyanopropionate
Formel: C₆H₉NO₂
Molecular Weight: 127,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013812
CAS-Nummer: 1572-99-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR