Norcampher 95%

Lieferant: Apollo Scientific

Synonyme: 2,5-Methanocyclohexanon, 2-Norbornanon, 2-Oxonorbornan, Bicyclo[2.2.1]-2-heptanon

OR921159-25G
APOSOR921159-25GEA 82.8 EUR
APOSOR921159-25G
Norcampher 95%
Norcampher
Formel: C₇H₁₀O
Molecular Weight: 110,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00074823
CAS-Nummer: 497-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR