4-Chlorothiophene-2-methanol 95%

Lieferant: Apollo Scientific

OR921216-1G OR921216-250MG
APOSOR921216-1GEA 806 EUR
APOSOR921216-1G APOSOR921216-250MG
4-Chlorothiophene-2-methanol 95%
4-Chlorothiophene-2-methanol
Formel: C₅H₅ClOS
Lagertemperatur: Raumtemperatur
MDL: MFCD17016116
CAS-Nummer: 233280-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR