1-Phenylpyrrol 95%

Lieferant: Apollo Scientific

Synonyme: N-Phenylpyrrol

OR921374-25G
APOSOR921374-25GEA 125 EUR
APOSOR921374-25G
1-Phenylpyrrol 95%
1-Phenylpyrrol
Formel: C₁₀H₉N
Molecular Weight: 143.19 g/mol
Siedepunkt: 234
Lagertemperatur: Raumtemperatur
MDL: MFCD00005343
CAS-Nummer: 635-90-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR