4-Isopropoxy-2-butanon 95%

Lieferant: Apollo Scientific

OR921450-10G OR921450-1G OR921450-5G
APOSOR921450-10GEA 632 EUR
APOSOR921450-10G APOSOR921450-1G APOSOR921450-5G
4-Isopropoxy-2-butanon 95%
4-Isopropoxy-2-butanon
Formel: C₇H₁₄O₂
Molecular Weight: 130,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059358
CAS-Nummer: 32541-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR