Bicyclo[4.1.0]heptan-2-one
Lieferant: Apollo Scientific
OR921618-1G
OR921618-250MG
OR921618-5G
APOSOR921618-1GEA
397
EUR
APOSOR921618-1G
APOSOR921618-250MG
APOSOR921618-5G
Bicyclo[4.1.0]heptan-2-one
Bicyclo[4.1.0]heptan-2-one
Formel:
C₇H₁₀O Molecular Weight: 110,16 g/mol Siedepunkt: 91 °C/15mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD09866571 CAS-Nummer: 5771-58-4 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...