2-(Methylsulphonyl)thiophene 95%

Lieferant: Apollo Scientific

OR921801-5G
APOSOR921801-5GEA 154 EUR
APOSOR921801-5G
2-(Methylsulphonyl)thiophene 95%
2-(Methylsulphonyl)thiophene
Formel: C₅H₆O₂S₂
Molecular Weight: 162.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052826
CAS-Nummer: 38695-60-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR