2-(4-Methylpiperazin-1-ylmethyl)phenylamine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-((4-Methylpiperazin-1-yl)methyl)aniline, 2-[(4-Methylpiperazin-1-yl)methyl]aniline
OR9219-1G
OR9219-250MG
APOSOR9219-1GEA
373
EUR
APOSOR9219-1G
APOSOR9219-250MG
2-(4-Methylpiperazin-1-ylmethyl)phenylamine 97%
2-(4-Methylpiperazin-1-ylmethyl)phenylamine
Formel:
C₁₂H₁₉N₃ Molecular Weight: 205.31 g/mol Schmelzpunkt: 82…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06804544 CAS-Nummer: 19577-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...