4-Nitrobenzyl-acetat 95%

Lieferant: Apollo Scientific

Synonyme: Acetic Acid 4-Nitrobenzyl Ester

OR922040-25G OR922040-5G
APOSOR922040-25GEA 150 EUR
APOSOR922040-25G APOSOR922040-5G
4-Nitrobenzyl-acetat 95%
4-Nitrobenzyl-acetat
Formel: C₉H₉NO₄
Molecular Weight: 195,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00024781
CAS-Nummer: 619-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR