N-Hydroxy-1-naphthamid 95%

Lieferant: Apollo Scientific

Synonyme: N-Hydroxynaphthalene-1-carboxamide, N-Hydroxy-1-naphthamide

OR922061-1G OR922061-250MG
APOSOR922061-1GEA 250 EUR
APOSOR922061-1G APOSOR922061-250MG
N-Hydroxy-1-naphthamid 95%
1-Naphthohydroxamic acid
Formel: C₁₁H₉NO₂
Molecular Weight: 187.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059546
CAS-Nummer: 6953-61-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR