3-Buten-1-yl-acetat 95%

Lieferant: Apollo Scientific

Synonyme: Acetic Acid 3-Butenyl Ester, 4-Acetoxy-1-butene

OR922099-25G OR922099-5G
APOSOR922099-25GEA 429 EUR
APOSOR922099-25G APOSOR922099-5G
3-Buten-1-yl-acetat 95%
3-Buten-1-yl-acetat
Formel: C₆H₁₀O₂
Molecular Weight: 114,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00086596
CAS-Nummer: 1576-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR