Di(2-pyridyl)ketonoxim 95%

Lieferant: Apollo Scientific

Synonyme: 2,2'-Dipyridyl ketoxime

OR922157-1G OR922157-5G
APOSOR922157-1GEA 219 EUR
APOSOR922157-1G APOSOR922157-5G
Di(2-pyridyl)ketonoxim 95%
Di(2-pyridyl)ketonoxim
Formel: C₁₁H₉N₃O
Molecular Weight: 199,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006311
CAS-Nummer: 1562-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR