1-Methyl-1H-indole-4-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methyl-4-indolecarboxylic Acid
OR9222-1G
OR9222-5G
OR9222-250MG
APOSOR9222-1GEA
58
EUR
APOSOR9222-1G
APOSOR9222-5G
APOSOR9222-250MG
1-Methyl-1H-indole-4-carboxylic acid 95%
1-Methyl-1H-indole-4-carboxylic acid
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol Schmelzpunkt: 243…245 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08435874 CAS-Nummer: 90924-06-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...