Isobutamben (Isobutyl-4-aminobenzoat) 98%

Lieferant: Apollo Scientific

Synonyme: 4-Aminobenzoic acid isobutyl ester

OR922243-1G OR922243-25G OR922243-5G
APOSOR922243-1GEA 21.9 EUR
APOSOR922243-1G APOSOR922243-25G APOSOR922243-5G
Isobutamben (Isobutyl-4-aminobenzoat) 98%
Isobutamben (Isobutyl-4-aminobenzoat)
Formel: C₁₁H₁₅NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD00017110
CAS-Nummer: 94-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR