4-Chlor-2-Methylbutyric Säure Methylester 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-methylbutyric Acid Methyl Ester

OR922351-5G
APOSOR922351-5GEA 197 EUR
APOSOR922351-5G
4-Chlor-2-Methylbutyric Säure Methylester 95%
4-Chlor-2-Methylbutyric Säure Methylester
Formel: C₆H₁₁ClO₂
Molecular Weight: 150,60 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00198001
CAS-Nummer: 13888-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR