Ethyl(methylthio)acetat 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl(methylmercapto)acetat, (Methylmercapto)essigsäureethylester, (Methylthio)essigsäureethylester

OR922368-100G
APOSOR922368-100GEA 68.3 EUR
APOSOR922368-100G
Ethyl(methylthio)acetat 95%
Ethyl(methylthio)acetat
Formel: C₅H₁₀O₂S
Molecular Weight: 134,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009182
CAS-Nummer: 4455-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR