Isovalerophenon 95%
Lieferant: Apollo Scientific
Synonyme:
3-Methyl-1-phenyl-1-butanon
OR922452-25G
OR922452-5G
APOSOR922452-25GEA
278
EUR
APOSOR922452-25G
APOSOR922452-5G
Isovalerophenon 95%
Isovalerophenon
Formel:
(C₆H₅)C(O)CH₂CH(CH₃)CH₃ Molecular Weight: 162,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00026486 CAS-Nummer: 582-62-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...