2-Ethylhexylamin 95%
Lieferant: Apollo Scientific
Synonyme:
1-Amino-2-ethylhexan
OR922474-100G
OR922474-1KG
OR922474-500G
APOSOR922474-100GEA
125
EUR
APOSOR922474-100G
APOSOR922474-1KG
APOSOR922474-500G
2-Ethylhexylamin 95%
2-Ethylhexylamin
Formel:
C₈H₁₉N Molecular Weight: 129,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008148 CAS-Nummer: 104-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...