Desoxybenzoin 97%

Lieferant: Apollo Scientific

Synonyme: 2-Phenylacetophenone, 1,2-Diphenylethanone

OR922691-500G
APOSOR922691-500GEA 127 EUR
APOSOR922691-500G
Desoxybenzoin 97%
Desoxybenzoin
Formel: C₁₄H₁₂O
Molecular Weight: 196,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003081
CAS-Nummer: 451-40-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR