Isopropyl crotonate 95%
Lieferant: Apollo Scientific
Synonyme:
Crotonic Acid Isopropyl Ester, Isopropyl Crotonate
OR922692-100G
OR922692-25G
OR922692-5G
APOSOR922692-100GEA
574
EUR
APOSOR922692-100G
APOSOR922692-25G
APOSOR922692-5G
Isopropyl crotonate 95%
Isopropyl crotonate
Formel:
C₇H₁₂O₂ Molecular Weight: 128,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059053 CAS-Nummer: 6284-46-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...