3-Methoxydiphenylamin 98%

Lieferant: Apollo Scientific

Synonyme: 3-Methoxy-N-phenylaniline

OR922698-100G OR922698-500G
APOSOR922698-100GEA 68.3 EUR
APOSOR922698-100G APOSOR922698-500G
3-Methoxydiphenylamin 98%
3-Methoxydiphenylamin
Formel: C₁₃H₁₃NO
Molecular Weight: 199,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008383
CAS-Nummer: 101-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR