4-Chloropyrido[2,3-d]pyrimidine 95%
Lieferant: Apollo Scientific
OR922751-100MG
OR922751-1G
OR922751-250MG
APOSOR922751-100MGEA
200
EUR
APOSOR922751-100MG
APOSOR922751-1G
APOSOR922751-250MG
4-Chloropyrido[2,3-d]pyrimidine 95%
4-Chloropyrido[2,3-d]pyrimidine
Formel:
C₇H₄ClN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06254345 CAS-Nummer: 28732-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...