Ethyl-tert-Butylacetat 96%

Lieferant: Apollo Scientific

Synonyme: tert-Butylacetic Acid Ethyl Ester, Ethyltert-butylacetate, 3,3-Dimethylbutyric Acid Ethyl Ester, Ethyl 3,3-Dimethylbutyrate

OR922819-100G OR922819-25G
APOSOR922819-100GEA 188 EUR
APOSOR922819-100G APOSOR922819-25G
Ethyl-tert-Butylacetat 96%
Ethyl-tert-butylacetate
Formel: C₈H₁₆O₂
Molecular Weight: 144.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00026911
CAS-Nummer: 5340-78-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR