Ethyl-4-chlor-1-piperidincarboxylat 95%
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-1-(ethoxycarbonyl)piperidine, 1-Carbethoxy-4-chloropiperidine, 4-Chloro-1-piperidinecarboxylic Acid Ethyl Ester, Ethyl 4-chloropiperidine-1-carboxylate
OR922822-5G
APOSOR922822-5GEA
92.8
EUR
APOSOR922822-5G
Ethyl-4-chlor-1-piperidincarboxylat 95%
Ethyl-4-chloro-1-piperidinecarboxylate
Formel:
C₈H₁₄ClNO₂ Molecular Weight: 191,66 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09038524 CAS-Nummer: 152820-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...